missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Caps with PTFE-Faced Foamed Polyethylene Liners

Product Code. 11754229
Click to view available options
Neck Finish:
20-400
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
89-400
Closure Liner:
PTFE/Polyethylene
Polyethylene Foam
Material:
Polypropylene
No. per Case:
144 Cs.
48 Cs.
72 Cs.
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Excellent for environmental samples, pharmaceuticals and diagnostic reagents

  • PTFE faced foamed polyethylene liner offers chemical resistance
  • Polyethylene foam provides compressibility and sealing properties
  • Good for high purity chemicals, strong acids, and solvents
  • For usewith glass bottles or vials
  • Polypropylene
  • Screw cap
  • White

TRUSTED_SUSTAINABILITY

Spécification

Color White
Material Polypropylene
No. per Case 48 Cs.
Neck Finish 70-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Glass bottle or vial
Closure Liner PTFE/Polyethylene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'