missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Caps with PTFE-Faced Foamed Polyethylene Liners

Product Code. 11784219
Click to view available options
Neck Finish:
20-400
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
89-400
Closure Liner:
PTFE/Polyethylene
Polyethylene Foam
Material:
Polypropylene
No. per Case:
144 Cs.
48 Cs.
72 Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

Excellent for environmental samples, pharmaceuticals and diagnostic reagents

  • PTFE faced foamed polyethylene liner offers chemical resistance
  • Polyethylene foam provides compressibility and sealing properties
  • Good for high purity chemicals, strong acids, and solvents
  • For usewith glass bottles or vials
  • Polypropylene
  • Screw cap
  • White

TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Polypropylene
No. per Case 144 Cs.
Neck Finish 20-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Glass bottle or vial
Closure Liner PTFE/Polyethylene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'