missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Open-Top Caps w/PTFE Faced Silicone Liners for E-Z Ex-Traction™ Vials

Suitable for storage and containment of volatile solvents, sensitive compounds or corrosive chemicals

242.00 € - 1135.00 €

Specifications

Color White
Material Polypropylene
Certifications/Compliance ISO 9001
Autoclavable Autoclavable
For Use With (Equipment) Glass bottle or vial
View More Specs

Products 8
Product Code Brand Neck Finish No. per Case Price Quantity & Availability  
Product Code Brand Neck Finish No. per Case Price Quantity & Availability  
10725373
View Documents
DWK Life Sciences
W240840
13-425 250 Cs.
291.00 €
Pack of 250
Estimated Shipment: 16-07-2025
to see stock.
Add to basket
 
10526863
View Documents
DWK Life Sciences
W240841
13-425 1000 Cs.
1050.00 €
Pack of 1000
Estimated Shipment: 26-03-2025
to see stock.
Add to basket
 
10048272
View Documents
DWK Life Sciences
W240842
15-425 250 Cs.
305.00 €
Pack of 250
Estimated Shipment: 31-03-2025
to see stock.
Add to basket
 
10139822
View Documents
DWK Life Sciences
W240843
15-425 1000 Cs.
1135.00 €
Pack of 1000
Estimated Shipment: 26-03-2025
to see stock.
Add to basket
 
10556863
View Documents
DWK Life Sciences
W240844
22-400 100 Cs.
253.00 €
Pack of 100
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
 
10119922
View Documents
DWK Life Sciences
W240845
22-400 500 Cs.
1085.00 €
Pack of 500
Estimated Shipment: 26-03-2025
to see stock.
Add to basket
 
10785553
missing translation for 'viewDocuments'
DWK Life Sciences
W240846
24-400 100 Cs.
242.00 €
Pack of 100
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
 
10557833
missing translation for 'viewDocuments'
DWK Life Sciences
W240847
24-400 500 Cs.
1120.00 €
Pack of 500
Estimated Shipment: 16-07-2025
to see stock.
Add to basket
 
Description

Description

Suitable for storage and containment of volatile solvents, sensitive compounds or corrosive chemicals

  • Autoclavable

Specifications

Specifications

White
ISO 9001
Glass bottle or vial
Polypropylene
Autoclavable
PTFE/Silicone
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.