missing translation for 'onlineSavingsMsg'
Learn More

Nasco Whirl-Pak™ Speci-Sponge™ Environmental Surface Sampling Bags
GREENER_CHOICE

Convenient sterile absorbent sponge sampler in a sterile Whirl-Pak bag.

Brand:  Nasco B01299WA

 View more versions of this product

Product Code. 11711806

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Convenient sterile absorbent sponge sampler in a sterile Whirl-Pak bag

  • Designed for testing work surfaces and equipment for Salmonella, Listeria, and other foodborne pathogens
  • Dry “Speci-Sponge” measures 1.5 × 3 × 0.63 in. (3.8 × 7.6 × 1.6cm); not pretreated with bactericide
  • Simply add diluent, wipe test surface, and return sponge to its 18 oz. (532mL) bag
  • Self-contained sample can then be shipped for analysis
  • Sponge will wipe a large area; bag's puncture-proof tabs reduce damage to sample
  • White write-on area allows easy labeling and identification

Order Info

Available with or without sterile glove.

Specifications

Specifications

With Dry Sponge
1.6 L
0.102 mm
12 x 7.5 in. (30 x 19 cm)
30 cm
19 cm
55 oz.
For environmental surfaces
4 mil
12 in.
7.5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.