missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ White 96-Well Immuno Plates

Product Code. 10396181
Click to view available options
Well Shape:
Round, Flat Bottom (F-well)
Surface Treatment:
MaxiSorp
Microfluor 1
Microfluor 2
Microlite 1+
Microlite 2+
PolySorp
Universal Binding
Volume (Metric) Working:
280 μL
330 μL
350 μL
Unit Size:
Pack of 50
Pack of 80

Product Code. 10396181

Brand: Thermo Scientific™ 436110

Only null left
Add to basket
Edge
Add to basket

Conduct quantitative and qualitative immunoassays like ELISA and binding assays with these white, 96-well immuno plates.

Conduct quantitative and qualitative immunoassays, like ELISA and binding assays, with Thermo Scientific™ White 96-Well Immuno Plates. These plates, which are compatible with common instruments, deliver optimal results, lot-to-lot reliability and well-to-well reproducibility.

Features

  • Plate has SBS footprint — fits standard equipment
  • Available with flat- or round-bottom wells
  • Choice of three surface types for optimal binding
  • Certified binding homogeneity/reproducibility

Choice of Surface:

  • Thermo Scientific PolySorp, Microfluor 1, Microlite 1+ and Universal Binding (UB) for adsorption of hydrophobic molecules
  • Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays
  • Thermo Scientific Microfluor 2 and Microlite 2+ are slightly hydrophilic and bind a diverse range of biomolecules

White:

  • Gives maximum reflection
  • Minimum autofluorescence and autoluminescence

Compatible with:

Lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane

Ordering Information:

Availability may vary by country.

TRUSTED_SUSTAINABILITY

Specifications

Binding Property Hydrophilic
Format Standard
Lid Without Lid
Detection Method Fluorescence
Array 8 x 12
Packaging 80 Cases
Color White
Product Type Microplate
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 350 μL
Sterility Non-sterile
Surface Treatment MaxiSorp
Well Shape Round, Flat Bottom (F-well)
For Use With (Application) Fluorescence and Luminescence
No. per Case 80
No. per Pack 10
Quantity 80/Cs.
Target Molecule Antibodies (Glycoproteins)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.