Learn More
Thermo Scientific™ White 96-Well Immuno Plates
Conduct quantitative and qualitative immunoassays like ELISA and binding assays with these white, 96-well immuno plates.
Brand: Thermo Scientific™ 9502887
Description
Conduct quantitative and qualitative immunoassays, like ELISA and binding assays, with Thermo Scientific™ White 96-Well Immuno Plates. These plates, which are compatible with common instruments, deliver optimal results, lot-to-lot reliability and well-to-well reproducibility.
Features
- Plate has SBS footprint — fits standard equipment
- Available with flat- or round-bottom wells
- Choice of three surface types for optimal binding
- Certified binding homogeneity/reproducibility
Choice of Surface:
- Thermo Scientific PolySorp, Microfluor 1, Microlite 1+ and Universal Binding (UB) for adsorption of hydrophobic molecules
- Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays
- Thermo Scientific Microfluor 2 and Microlite 2+ are slightly hydrophilic and bind a diverse range of biomolecules
White:
- Gives maximum reflection
- Minimum autofluorescence and autoluminescence
Compatible with:
Lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane
Ordering Information:
Availability may vary by country.
Specifications
Hydrophobic | |
With Lid | |
8 x 12 | |
White | |
Plastic | |
330 μL | |
Universal Binding | |
Luminescence | |
25 | |
50/Cs. |
Standard | |
Chemiluminescence | |
50 Cases, 25 Pk. | |
Microplate | |
96 | |
Non-sterile | |
Round, Flat Bottom (F-well) | |
50 | |
None | |
Lipids |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.