Learn More
Thermo Scientific™ White and Black 384-Well Immuno Plates
GREENER_CHOICE
Conduct quantitative and qualitative immunoassays with these white and black 384-well immuno plates.
Brand:  Thermo Scientific™ 460372
295.94 EUR valid until 2024-12-31
Use promo code "21894" to get your promotional price.
 View more versions of this product
Product Code. 10385991
  • 394.00 € / Pack of 30
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Conduct quantitative and qualitative immunoassays with Thermo Scientific™ White and Black 384-Well Immuno Plates. Ideal for ELISA and binding assays, these plates are compatible with common instruments. They offer lot-to-lot reliability and well-to-well reproducibility for optimal results.
Features:
  • Plate has SBS footprint, fits standard equipment and is available with flat-bottom wells
  • Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays
  • Plate is offered as black or white for optimal choice of application type
  • Certified binding offers homogeneity/reproducibility.
White:
  • Maximum reflection/minimum autofluorescence and autoluminescence
Black:
  • Minimizes crosstalk, back-scattered light and background in fluorescence reading
Compatible with:
Lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane
Ordering information:
Availability may vary by country
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hydrophilic
Without Lid
16 x 24
White
Polystyrene
100 μL
MaxiSorp
Fluorescence and Luminescence
10
Antibodies (Glycoproteins)
Standard
Chemiluminescence
30 Cases
Microplate
384
Non-sterile
Square, Flat Bottom (F-well)
30
None
F384