missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ White and Black 384-Well Immuno Plates
Conduct quantitative and qualitative immunoassays with these white and black 384-well immuno plates.
Brand: Thermo Scientific™ 460518
239.36 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Conduct quantitative and qualitative immunoassays with Thermo Scientific™ White and Black 384-Well Immuno Plates. Ideal for ELISA and binding assays, these plates are compatible with common instruments. They offer lot-to-lot reliability and well-to-well reproducibility for optimal results.
Features:
- Plate has SBS footprint, fits standard equipment and is available with flat-bottom wells
- Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays
- Plate is offered as black or white for optimal choice of application type
- Certified binding offers homogeneity/reproducibility.
White:
- Maximum reflection/minimum autofluorescence and autoluminescence
Black:
- Minimizes crosstalk, back-scattered light and background in fluorescence reading
Compatible with:
Lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane
Ordering information:
Availability may vary by country
Specifications
Hydrophilic | |
Without Lid | |
16 x 24 | |
Black | |
Polystyrene | |
100 μL | |
MaxiSorp | |
Fluorescence | |
10 | |
Antibodies (Glycoproteins) |
Standard | |
Fluorescence | |
30 Cases, 10 Pk. | |
Microplate | |
384 | |
Non-sterile | |
Square, Flat Bottom (F-well) | |
30 | |
None | |
F384 |