missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ White Coverall
BIOPHARMA

Product Code. 15248128
This item is not returnable. View return policy
This item is not returnable. View return policy

The BioClean-D™ coverall with hood is a disposable garment featuring a front zip with protective flap, elasticated hood, back, cuffs and ankles and thumb loops to ensure a secure hold. The BioClean-D coverall provides exceptional comfort and protection, even when handling cytotoxic drugs

Cuffs & ankles ISO Class 4 cleanroom compatible zip with adhesive cover flap, Type 5 & 6 White XL BioClean-D
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size X-Large
Color White
Disposable Single-use
Sterility Non-sterile
Size (Metric) Chest 108 to 116 cm
For Use With (Application) Controlled Environments
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.