Learn More
3M™ White Disposable Visitor Lab Coat, Series 4400
Offers a barrier against non-hazardous particles and liquid splashes. The Series 4400 Disposable Protective Lab Coat is anti-static treated and can be used in light duty applications.
123.42 € - 340.00 €
Specifications
Color White
Cuff Style Elastic
View More Specs
Products 9
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12857310
View Documents
3M™
7000089699
4X-Large
332.00 €
Pack of 50
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
12951335
View Documents
3M™
7000089695
Medium
340.00 €
Pack of 50
In Stock
to see stock.
Edge
Add to basket
12931335
View Documents
3M™
7000034776
Small This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12847310
View Documents
3M™
7000089698
3X-Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12921335
View Documents
3M™
7000089697
2X-Large
340.00 €
Pack of 50
In Stock
to see stock.
Add to basket
12981325
View Documents
3M™
7000034777
Large
340.00 €
Pack of 50
In Stock
to see stock.
Edge
Add to basket
12901335
View Documents
3M™
7000089696
X-Large
340.00 €
Pack of 50
In Stock
to see stock.
Edge
Add to basket
12941335
View Documents
3M™
7000034776
Small
332.00 €
Pack of 50
In Stock
to see stock.
Add to basket
15351495
View Documents
3M™
7000089698
3X-Large
332.00 €
Pack of 50
Estimated Shipment: 27-03-2025
to see stock.
Add to basket
Description
Description
  • Breathable, lightweight material.
  • Designed for light duty applications.
  • Offers a barrier against non-hazardous particles and liquid splashes.
  • Zip fastener.
  • Anti-static treated.
  • Elastic cuffs
Specifications
Specifications
White
Elastic