Learn More
Moldex™ White Face Mask
Extremely low breathing resistance increases the performance of both mask and wearer
Brand:  Moldex™ 320501
Product Code. 12560237
  • 173.00 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
The uniquely developed Series 3000 Air from Moldex offers extremely low breathing resistance and therefore increased performance of both mask and wearer. The filtering surface is almost three times larger than a standard filter product, allowing you to breathe more easily.
Specifications
Specifications
EN 149:2001A1:2009
Single-use
Face Mask
Protects against dust, mist and fumes
30 g
White
Filter Layer, Inner Shell, DuraMesh: Polypropylene, Ethylene-vinyl acetate (EVA), Nose Seal, Clip: Polyethylene, Head Strap: Polyester, Lycra, Ventex-valve: Natural Rubber
Medium/Large
10/Box