missing translation for 'onlineSavingsMsg'
Learn More

3M™ White Disposable Polypropylene Coveralls, 4500 Series

Product Code. 13463686
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Height (English):
64 to 67 in.
66 to 69 in.
69 to 71 in.
70 to 74 in.
73 to 76 in.
76 to 78 in.
78 to 81 in.
Height (Metric):
164 to 170 cm
167 to 176 cm
174 to 181 cm
179 to 187 cm
186 to 194 cm
194 to 200 cm
200 to 206 cm
Unit Size:
Pack of 20
This item is not returnable. View return policy

Product Code. 13463686

Brand: 3M™ 7000089592

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Disposable polypropylene coveralls offer Category 1 protection in non-hazardous applications

CE Simple, Category 1 coverall is designed for domestic DIY use, light-duty industrial cleaning and maintenance applications, or environments where hygiene and cleanliness are important.
  • Excellent economic solution to keep the wearer clean and dry
  • Suitable for non-hazardous applications
  • Breathable and light-duty protection
  • Strong, durable 45 gsm polypropylene fabric
  • Storm flap over zip
  • Elasticated hood, cuffs, ankles, and waist
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protective Coveralls
Size Large
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Hood Type 2-panel
Closure Type Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Protection Class Category I
Model 4500W
Size (English) Chest 39 to 43 in.
Size (Metric) Chest 100 to 108 cm
For Use With (Application) For domestic DIY use, light duty industrial cleaning and maintenance applications or environments
Height (English) 69 to 71 in.
Height (Metric) 174 to 181 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt