missing translation for 'onlineSavingsMsg'
Learn More

3M™ White Disposable Polypropylene Coveralls, 4500 Series

Product Code. 13473686
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Height (English):
64 to 67 in.
66 to 69 in.
69 to 71 in.
70 to 74 in.
73 to 76 in.
76 to 78 in.
78 to 81 in.
Height (Metric):
164 to 170 cm
167 to 176 cm
174 to 181 cm
179 to 187 cm
186 to 194 cm
194 to 200 cm
200 to 206 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

Use for non-hazardous applications. The White Disposable Coveralls, 4500 Series offers Category I protection from polypropylene fabric.

CE Simple, Category 1 coverall is designed for domestic DIY use, light-duty industrial cleaning and maintenance applications, or environments where hygiene and cleanliness are important.
  • Excellent economic solution to keep the wearer clean and dry
  • Suitable for non-hazardous applications
  • Breathable and light-duty protection
  • Strong, durable 45gsm polypropylene fabric
  • Storm flap over zip
  • Elasticated hood, cuffs, ankles, and waist
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protective Coveralls
Size X-Large
Material Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Hood Type 2-panel
Closure Type Sealed Storm Flap
Seam Type Serged
Protection Class Category I
Size (English) Chest 43 to 45 in.
Size (Metric) Chest 108 to 115 cm
For Use With (Application) For domestic DIY use, light duty industrial cleaning and maintenance applications or environments
Height (English) 70 to 74 in.
Height (Metric) 179 to 187 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'