missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ White Urea Screw Caps

Suitable for most applications that call for screw neck bottles. Wheaton™ White Urea Screw Caps are EPE lined.

Brand:  Wheaton™ 300190

112.10 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 14813592

  • 125.00 € / Pack of 1000
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Multiple thread size options
  • Excellent closing option for screw neck bottles and vials
TRUSTED_SUSTAINABILITY
Specifications

Specifications

15.5 mm
Cap
White
Screw Thread Diagnostic Vials
EPE
Urea
1000 Pack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers