missing translation for 'onlineSavingsMsg'
Learn More

Brannan™ White Wall Thermometer

Takes measurements with miscible red spirit filling. Brannan™ White Wall Thermometer. Displays both °C and °F temperature scales and is suitable for indoor and outdoor use.

Brand:  Brannan™ 14/435/3

Product Code. 13068623

  • 4.60 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Takes measurements with miscible red spirit filling. Brannan™ White Wall Thermometer. Displays both °C and °F temperature scales and is suitable for indoor and outdoor use.

Plastic wall thermometer with black printed temperature scale on white moulding.
  • Suitable for indoor and outdoor use
  • Displays both °C and °F temperature scales
  • Filled with miscible red spirit filling
  • Length: 215mm
Specifications

Specifications

295 x 85 x 0.5 mm
Styrene
22 g
120°F
0°F
0°F to 120°F
±1.0°C at ambient
250 mm
Red Spirit
Standard Wall Thermometer
50°C
-20°C
-20°C to +50°C
1°C/2°F
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.