missing translation for 'onlineSavingsMsg'
Learn More

Manutan Whiteboard, Ekwo, lacquered steel, 1200mm x 1800mm

Brand:  Manutan A337767

Encompass_Preferred

Product Code. 12867418

  • 261.72 € / Each
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

White, pre-lacquered, magnetic steel back-board. Jottings can be erased when dry with pen tray below the board. Elegant, smooth, anodised aluminum framework with polypropylene rounded corners. Invisible fixing points with screws and plugs supplied.
Dimensions (H x W) : 1200mm x 1800mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers