missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Wide-field Microscope Eyepiece, 13mm
Use this eyepiece with Steddy Trinocular Stereo Microscopes. Fisherbrand™ Wide-field Microscope Eyepieces are ergonomically designed for everyday use.
Brand: Fisherbrand™ 7500.0015
This item is not returnable.
View return policy
Description
- For Steddy trinocular stereo microscope
Specifications
Widefield Eyepiece | |
13 mm | |
Steddy trinocular stereo microscope | |
15X |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction