missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Wide Neck Conical Borosilicate Glass Erlenmeyer Flasks

Constructed from borosilicate glass. Fisherbrand™ Wide Neck Conical Borosilicate Glass Erlenmeyer Flasks are graduated and available in a variety of sizes.

Brand:  Fisherbrand™ SPFS66650-300

51.86 EUR valid until 2024-11-30
Use promo code "21756" to get your promotional price.


 View more versions of this product

Product Code. 15419113

  • 87.65 € / Pack of 10
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Borosilicate glass
  • Graduated
TRUSTED_SUSTAINABILITY
Specifications

Specifications

50 mm
Borosilicate Glass
Yes
300 mL
156 mm
87 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers