Learn More
Fisherbrand™ Wide Neck Conical Borosilicate Glass Erlenmeyer Flasks
Constructed from borosilicate glass. Fisherbrand™ Wide Neck Conical Borosilicate Glass Erlenmeyer Flasks are graduated and available in a variety of sizes.
Brand:  Fisherbrand™ SPFS66650-1000
108.70 EUR valid until 2024-11-30
Use promo code "21756" to get your promotional price.
 View more versions of this product
Product Code. 15466133
  • 167.00 € / Pack of 10
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Borosilicate glass
  • Graduated
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DIN ISO 24450
1 L
Wide neck
Yes