missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Wide Neck Conical Borosilicate Glass Erlenmeyer Flasks
Constructed from borosilicate glass. Fisherbrand™ Wide Neck Conical Borosilicate Glass Erlenmeyer Flasks are graduated and available in a variety of sizes.
Brand: Fisherbrand™ SPFS66650-50
43.49 EUR valid until 2024-11-30
Use promo code "21756" to get your promotional price.
Description
- Borosilicate glass
- Graduated
Specifications
34 mm | |
Borosilicate Glass | |
Yes |
50 mL | |
85 mm | |
51 mm |