Learn More
Cole-Parmer™ Wildco™ Kemmerer Water Sampler
Sample at specific depths between 3 and 600 feet.
Brand:  Cole-Parmer™ 05485-00
 View more versions of this product
Product Code. 11774499
  • 964.00 € / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • WildcoKemmerer Water Sampler
  • The messenger activates the unique trip heads that ensure closure in fast flowing streams or turbulent waters, regardless of line angle
  • All models include a plastic carrying case; order messenger and line separately.
  • The 304 SS models have urethane end seals (do not use when mercury concentrations exceed 1 ppm).
Specifications
Specifications
Kemmerer Water Sampler
Stainless Steel
3 to 600 ft depth
400 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products