missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ William's E Medium, no glutamine
GREENER_CHOICE

William's E Medium was originally developed by Williams and Gunn as reduced serum-supplemented medium for long-term cell cultures of adult rat liver epithelial cells.

Brand:  Gibco™ 12551032

 View more versions of this product

Product Code. 12599059

  • 44.83 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

William's E Medium was originally developed by Williams and Gunn as reduced serum-supplemented medium for long-term cell cultures of adult rat liver epithelial cells. Gibco™ William's E Medium can also be used for growing hepatocyte cells of different species, such as human derived HepaRG™ cells.

This William's E Medium is modified as follows:

  • With: Phenol Red
  • Without: L-glutamine, HEPES

The complete formulation is available.

Gibco™ William's E Medium is a modification of earlier media that has been enriched in amino acids and double the glucose. William's E Medium contains unique ingredients, including zinc, iron, manganese, non-essential amino acids, the reducing agent glutathione and the lipid methyl linoleate.

Dual-Site cGMP Manufacturing

Gibco™ William's E Medium is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ William's E Medium product made in our Grand Island facility (12551-032). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

William's E Medium contains no proteins or growth factors. Therefore, William's E Medium requires supplementation, commonly with 5-10% Fetal Bovine Serum (FBS). William's E Medium uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Liquid
Phenol Red, Sodium Pyruvate
Hepatocytes and hepatic models, mammalian cell culture, pharma and biopharma, primary cell culture
Room Temperature
Mammalian Cell Culture
William's E Medium
No Glutamine, No HEPES
500 mL
Storage conditions: 2°C to 8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2428784 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12551032
Lot Number2537738 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12551032
Lot Number2187225 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12551032
Lot Number2393329 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12551032
Lot Number2393371 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12551032
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ William's E Medium, no glutamine > Quantity: 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.