Learn More
Gibco™ William's E Medium, no phenol red
William's E Medium was originally developed by Williams and Gunn as reduced serum-supplemented medium for long-term cell cultures of adult rat liver epithelial cells. Gibco™ William's E Medium can also be used for growing hepatocyte cells of different species, such as human derived HepaRG™ cells.
Brand: Gibco™ A1217601
42.63 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Description
Gibco™ William's E Medium is a modification of earlier media that has been enriched in amino acids and double the glucose. William's E Medium contains unique ingredients, including zinc, iron, manganese, non-essential amino acids, the reducing agent glutathione and the lipid methyl linoleate.
Dual-Site cGMP Manufacturing
For supply chain continuity, we manufacture Gibco™ William's E Medium at two separate facilities located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements, are certified to ISO 13485, and are registered with the FDA as medical device manufacturers.
William's E Medium contains no proteins or growth factors. Therefore, William's E Medium requires supplementation, commonly with 5-10% Fetal Bovine Serum (FBS). William's E Medium uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications
1 :1 | |
1 X | |
William's E Medium | |
Sodium Pyruvate | |
Sustainable packaging | |
Room Temperature |
Store in refrigerator (2–8°C) and protect from light. | |
Liquid | |
Sterile | |
No Glutamine, No HEPES, No Phenol Red | |
500 mL |
For Research Use Only. Not for use in diagnostic procedures.