missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Wilmad™ 3mm O.D. High-Throughput Thin Walled NMR Tubes, 4 in.L, With Bruker SampleJet Cap

Thoroughly inspected to ensure the success of your experiments - designed for routine use in most low- to mid-field NMR spectrometers.

Brand:  Bel-Art™ 66000-0077

Product Code. 15308658

  • 315.00 € / Pack of 100
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket

Alert:

Do not use with ceramic spinners.

This item is not returnable. View return policy

Description

Description

Sold 100 per pack
Specifications

Specifications

102 mm
High-Throughput
Tube & Bruker™ SampleJet Cap
0.27 mm
Wilmad™ WG-3000-4-SJ
2.99±0.03 mm
For use in Bruker™ SampleJet system.
4 in.
100/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.