missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Wilmad™ 4.0mm Solid State NMR Rotor Cap Remover

Eliminate possible damage of the cap and rotor with this zero turn cap remover. Bel-Art SP Scienceware™ Wilmad™ 4.0mm Solid State NMR Rotor Cap Remover simplifies removal of end caps and base plugs in solid state NMR.

Brand:  Bel-Art™ 66000-0041

Product Code. 15358608

  • 2925.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • For use with 4mm solid state caps
Specifications

Specifications

4 mm Rotor
Rotor Cap Remover
RS-301-4180
4.0mm Solid State NMR Rotor Cap Remover
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers