Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Aldrich FT-NMR Library with Spectral Viewer on CD-ROM
Allows for Spectra processing using Zoom, Peak Integration and Peak Labeling. The Bel-Art SP Scienceware™ Wilmad™ Aldrich FT-NMR Library with Spectral Viewer on CD-ROM is an electronic book that contains high-resolution 300MHz 1H and 75MHz 13C spectra, integral, chemical structure and technical data for the more than 11,800 compounds.
765.00 € - 1670.00 €
Specifications
Description FT-NMR Library with Spectral Viewer on CD-ROM
View More Specs
Products 2
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
15338698
View Documents
Bel-Art™
66000-0107
1,1800 Compound Database This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15328698
View Documents
Bel-Art™
66000-0106
3,500 Compound Database This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Supplemental library that includes spectra for 3,500 additional compounds not included in the original printed version is also available
  • Order both the standard and the supplemental library for access to spectra on over 15,000 compounds
  • Multiple search methods: text Search by Chemical Name, Aldrich Catalog Number, CAS Number, Molecular Formula, advanced data field search on chemical categories and physical properties, such as Boiling Point, Melting Point, Density, Flash Point, Molecular Weight and Refractive Index
  • Minimum System Requirement: Windows Platform, CD-ROM drive, 600MB hard disk space
Specifications
Specifications
FT-NMR Library with Spectral Viewer on CD-ROM