Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 4 mm Finger for Varian/Bruker
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 4 mm Finger for Varian/Bruker is manufactured to precision standards that exceed spectrometer products in terms of dimension tolerance, reliability and life span.
Brand: Bel-Art™ SP Scienceware™ 66000-0056
This item is not returnable.
View return policy
Description
- Cost effective compared to the full VT version for sample changers
- Allows a portion of the VT air to pass straight past the sample inside the spinner turbine therefore reducing VT gradients and micro sonic flutter due to a high VT flow
- Includes high temperature external O-rings at the top and bottom for long life and a firm grip on the sample at top and bottom.
- Can also be used for spinning experiments.
Specifications
Agilent/Varian NMR Spectrometer | |
UNI-FINGER-PEEK-4 |