missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Wilmad™ Low Pressure Vacuum Tube

Offers a convenient flame-free sealing solution for air sensitive or volatile liquid samples. Bel-Art SP Scienceware™ Wilmad™ Low pressure Vacuum Tube is ideal for anaerobic and gas-tight EPR experiments.

Brand:  Bel-Art™ 66504-0585

 View more versions of this product

Product Code. 15388988

  • 199.00 € / Each
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket

Alert:

Due to the nature of glass, Extreme Caution should be exercised when using at elevated or reduced pressures since a tiny scratch on the glass surface would significantly lower the tensile strength. Adequate safety shielding should always be used when working in these conditions.

This item is not returnable. View return policy

Description

Description

  • Robust sealing system allows pressure build-up inside the sample
  • Greaseless PTFE piston provides a 100% contamination-free seal
  • Redesigned with a 4X larger sealing surface, eliminating leaks and greatly increasing lifetime when compared to traditional J-Young tubes
  • Axial symmetric design guarantees application in spinning experiments
Specifications

Specifications

13 μm
203 mm
500MHz
8 in.
25μm
5 mm
Glass tube, piston, and vacuum adapter
528-LPV-8
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers