missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Precision Quartz EPR Sample Tube
- Hold 10 to 15 times better tolerances on the inner diameter and 5 times smaller camber and concentricity vs. economy brand tubes
- Ideal for continuous wave experiments that require highly reproducible sample volume in the cavity
- Individually wrapped to prevent contamination
Brand: Bel-Art™ 66000-0016
This item is not returnable.
View return policy
Description
- Hold 10 to 15 times better tolerances on the inner diameter and 5 times smaller camber and concentricity vs. economy brand tubes
- Ideal for continuous wave experiments that require highly reproducible sample volume in the cavity
- Individually wrapped to prevent contamination
Specifications
76 μm | |
250 mm | |
Sample Tube | |
3.20±0.013 mm | |
9.84 in. | |
715-PQ-250M |
15 μm | |
Quartz | |
4.00±0.013 mm | |
Cap | |
0.40 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction