Learn More
Bel-Art™ SP Scienceware™ Wilmad Rotor and Caps for Bruker MAS Probe
Eliminate the need to solvate with MAS rotors manufactured from the highest purity Zirconia. Bel-Art SP Scienceware™ Wilmad MAS Rotors provide the ultimate solution for analysis of solid samples. Available for most present solid state NMR spectrometers.
130.00 € - 7640.00 €
Products 7
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15388678
View Documents
Bel-Art™
66000-0096
One 2.5mm Rotor, Two Vespel Caps and Botoms
2520.00 €
Each
15398678
View Documents
Bel-Art™
66000-0097
Rotor Body, Both Ends Open
1255.00 €
Each
15318688
View Documents
Bel-Art™
66000-0098
One 3.2mm Rotor, Two Vespel Caps and Botoms
2745.00 €
Each
15328688
View Documents
Bel-Art™
66000-0099
Five Rotors, Ten Kel-F Caps and Three Torlon Caps
7640.00 €
Each
15338688
View Documents
Bel-Art™
66000-0100
Five Rotors, Ten Kel-F Caps and Five Torlon Caps
5760.00 €
Each
15348688
View Documents
Bel-Art™
66000-0101
Cap
130.00 €
Each
15378688
View Documents
Bel-Art™
66000-0102
Rotor Cap
331.00 €
Each
Description
Description
  • Caps fitted with O-rings for improved sealing
  • MAS Probe and caps are 100% compatible for most solid state NMR spectrometers
  • Thoroughly inspected before and after the precision machining process to ensure there are no material irregularities
  • Spin testing is performed to only the highest specified spinning speed, assuring performance without overspinning the rotor