Learn More
TFA Dostmann Wind Meter WIND BEE
The handy wind meter WIND BEE displays the current wind speed and the highest and lowest values in kilometers or miles per hour, in meters per second or in knots. In addition, the temperature is determined. WIND BEE has a tripod connector for measurements over a longer period of time.
Brand:  TFA Dostmann 42.6003.13
Product Code. 17931642
  • 72.30 € / Each
Explore more special offers
Description
Description
The handy anemometer WIND BEE by TFA is the ideal tool for all sailors, kite surfers and outdoor enthusiasts. WIND BEE is also a useful companion at work or in various leisure activities. The device displays the current wind speed and the highest and lowest values in kilometers or miles per hour, in meters per second or in knots. In addition, the temperature is determined. WIND BEE has a tripod connector for measurements over a longer period of time.
  • Ideal for easy measurement of wind speed (at work or during various leisure activities)
  • Highest and lowest values and display of the temperature
  • Accuracy ± 3%
  • Backlight
  • With tripod connector
Specifications
Specifications
Anemometer
±3%
Plastic