missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AZOWIPE™ Wipes
Bactericidal hard surface disinfectant wipes. AZOWIPE™ Wipes are convenient for use in all clinical environments and are an important part of a robust infection control protocol.
877.00 €
Product Code | Brand | Dimensions (L x W) | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Product Type | Price | Quantity & Availability | ||||
12889297
|
Vernacare
81160 |
220 x 200 mm | Wipes |
877.00 €
6kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Made from a viscose rayon nonwoven material strong and durable in use
- Foil sealed - prevents evaporation in storage
- Suitable for all hard surfaces - one product from a single source
- Contains NO preservative - no smearing or streaking when in use
- Specially designed canister - easy to dispense wipes
- 70% Solution of Isopropyl Alcohol BP
- AZOWIPE - for the disinfection of hard surfaces
- Dimensions: 200mm x 220mm
Specifications
Wipes | |
220 x 200 mm |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title