Learn More
Brady™ Wire and Cable Flag Polypropylene Labels
Easily flag cables and wires for maximum readability with thermal transfer, printable polypropylene flag label material (B-425). Print almost anywhere with a Brady™ portable label printer.
Brand:  Brady™ 170860
Product Code. 17881739
  • 49.65 € / Pack of 200
Explore more special offers
Description
Description
  • Designed to create pre-sized labels for flagging small diameter data cables and wires for increased visibility and easy removal
  • Resistant to extreme temperatures and solvents such as oils, chemicals and water
  • Made with a strong acrylic adhesive that remains tightly bonded
  • Meets requirements of a halogen-free material per DIN VDE 0472 Part 815
Specifications
Specifications
UL Recognized Component to UL969, CSA Accepted to C22.2 No. 0.15 to 95
Permanent Acrylic
Matte
Polypropylene
Non-sterile
BMP51, M511
-94°F to +266°F
1.575 x 1.181 in.
1.181 in.
M5-01-425-FT
Black on White
Die-cut
Labels
Asset Tracking, Barcode Labeling, Component and Equipment Labeling, Data and Telecommunications Labeling, General Identification, Inventory and Inspection Labeling, Laboratory Labeling, Wire and Cable Labeling
200 Labels/Cartridge
0.0046 in.
1.575 in.
3 cm