missing translation for 'onlineSavingsMsg'
Learn More

Manutan Wire rope strop, galvanised, 10mm diameter, 6x19 thimble eye, 2m long

Brand:  Manutan A341540

Encompass_Preferred

Product Code. 12831559

  • 34.49 € / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

10 x 19 construction wire rope to British Standards. Strops with eye each end. Stamped with serial number and WLL, safety Factor 5:1.
  • WLL (T): 1.0
  • Diameter: 10mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers