Learn More
Fisherbrand™ Wire Gauze
Provides excellent support to beakers and other glassware during heating on ring stands.Fisherbrand™ Wire Gauze is a flat square manufactured from galvanized iron mesh.
Brand:  Fisherbrand™ 9072
Product Code. 11789806
  • 28.65 € / Pack of 10
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Non-fraying edges
  • Durable and long-lasting
Specifications
Specifications
Wire Gauze