Learn More
Tocris Bioscience™ Withaferin A
Inhibits NF-κB activation
Brand:  Tocris Bioscience™ 2816/1
155.50 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18716627
  • 197.00 € / 1mg
Explore more special offers
Description
Description
Withaferin A is a steroid lactone that displays anti-inflammatory, antitumor and antiangiogenic activity. Withaferin A inhibits endothelial cells (HUVEC) sprouting in vitro (IC50 = 12 nM) and in vivo, prevents NF-κB activation by inhibiting activation of IKKβ and inhibits chymotrypsin-like activity of the 20S proteasome. In an animal model of Huntington's disease (HD), Withaferin A delays disease progression and increases lifespan; in a cellular model it suppresses aggregation of mutant huntingtin, activates HSF1 and induces Hsp70 chaperone expression.
Specifications
Specifications
>98%
1 mg
470.6
Store at -20°C
Withaferin A
Withaferin A