Learn More
Gibco™ Wnt Surrogate-Fc Fusion Recombinant Protein
Description
Gibco Wnt Surrogate-Fc Fusion Protein activates similar Wnt signaling pathways as most commercially available recombinant Wnt3a proteins or Wnt-conditioned media, while requiring a fraction of the amount of protein or hassle. Gibco Wnt Surrogate-Fc Fusion Protein was engineered with improved solubility, overcoming a major obstacle of working with Wnt3a and reducing the concentrations required to activate Wnt signaling in your model system. Gibco Wnt Surrogate-Fc Fusion Protein is comprised of the LRP-binding domain of DKK linked to a selective, high affinity binder of Frizzled receptors. Gibco Wnt Surrogate-Fc Fusion Protein is provided at a concentration of 2 mg/mL which is equivalent to 17.1 μM based on dimeric stoichiometry. Storage: Store undiluted Gibco Wnt Surrogate-Fc Fusion Protein at -80° for up to 1 year. After thawing, apportion into working aliquots and store at -20°C to -80°C for up to 2 months.
Specifications
Specifications
| Accession Number | P56704 |
| Concentration | 2 mg/mL |
| For Use With (Application) | Functional Assay |
| Formulation | 25 mM tris with 500 mM NaCl and no preservative; pH 8.2 |
| Gene ID (Entrez) | 89780 |
| Name | Wnt Surrogate-Fc Fusion |
| Quantity | 25 μg |
| Source | HEK293 cells |
| Regulatory Status | RUO |
| Endotoxin Concentration | 1 EU/mL |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.