missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microspec™ Wrappastack™ Sleeves
For storage of prepared petri dishes.
40.20 € - 43.35 €
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
15702145
|
Microspec™
K5 |
55 mm |
40.20 €
Pack of 200 |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15772135
|
Microspec™
K2 |
90 mm |
43.35 €
Pack of 200 |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
Description
- Professionally wrap poured plates for fridge storage or incubation
- Allows poured plates to heat shrunk into sleeves for fridge storage
- Keeps plates neatly stacked and steady
- Reduces moisture loss
- Tear strip for easy removal of plates when required
Specifications
Polypropylene | |
Sleeve | |
180 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title