missing translation for 'onlineSavingsMsg'
Learn More

Microspec™ Wrappastack™ Starter Kit

Complete kit for wrapping petri dishes

Brand:  Microspec™ K1

Product Code. 15762135

  • 210.00 € / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Petri Dish wrapping system
  • Allows poured plates to heat shrunk into sleeves for fridge storage or incubation
  • Keeps plates neatly stacked and steady
  • Reduces moisture loss
  • Tear strip for easy removal of plates

Includes: Heat Gun, Plate Stands and Sleeves for 90mm dishes

Specifications

Specifications

Polypropylene
Starter Kit
90 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.