missing translation for 'onlineSavingsMsg'
Learn More

Wright Stain, pure, certified, Thermo Scientific Chemicals

Product Code. 10537182
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68988-92-1
Molecular Formula C36H27Br4N3O5S+2
Molecular Weight (g/mol) 933.304
MDL Number MFCD00082143
InChI Key AXIKDPDWFVPGOD-UHFFFAOYSA-O
PubChem CID 25113599
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;2-(2,4,5,7-tetrabromo-3,6-dihydroxyxanthen-10-ium-9-yl)benzoic acid
SMILES CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.C1=CC=C(C(=C1)C2=C3C=C(C(=C(C3=[O+]C4=C(C(=C(C=C24)Br)O)Br)Br)O)Br)C(=O)O

Specifications

Solubility Information Solubility in water: insoluble.
Infrared Spectrum Authentic
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Green
Quantity 10 g
Physical Form Crystalline Powder
Chemical Name or Material Wright Stain, Pure

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'