missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark™ WypAll™ L40 Fabric Wipers
Use these wipers for industrial tasks such as cleaning, process line wiping and soaking up large spills. WypAll™ L40 Paper Wipers are made from a specialized fabric, featuring double re-creped (DRC) base sheet technology for balance of softness and strength.
Brand: Kimberly-Clark™ 7462
166.25 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- Large, perforated roll for use in busy areas
- Can be used with a dispensing system to help control use and reduce waste
Specifications
Medium-duty Industrial Wiper | |
25 cm | |
41 cm | |
White | |
A limited life wiper using double re-creped (DRC) base sheet technology, layered fabric that’s strong, thick and highly absorbent so it cleans up fast without falling apart, does the job with fewer wipers and helps reduce costs | |
Non-sterile |
9.8 in. | |
16.1 in. | |
Double Re-crepe Cellulose | |
25 x 41 cm (9.8 x 16.1 in.) | |
1-ply |