missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ WypAll™ X50 Cloths Large Roll, White

Specially designed for light-duty and intricate wiping tasks. WypAll™ Large Roll White X50 Cloths are ideal for use in manufacturing and food processing environments.

Brand:  Kimberly-Clark™ 8356

105.38 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.


Product Code. 12589727

  • 276.00 € / 1 roll
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Food Contact certified
  • Advanced HydroKnit™ Technology ensures each cloth is strong and fast absorbing
  • Designed for general purpose, light-duty and intricate wiping tasks
  • For use in manufacturing and food processing environments
  • Clean, reliably consistent and extended use cloths
  • Designed to help you improve productivity and efficiency whilst reducing waste
  • Suitable for hand wiping
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Light-duty Industrial Wiper
31 cm
24.9 cm
Food Contact
31 x 24.9 cm
Non-sterile
12.2 in.
9.8 in.
Paper
White
1-ply
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers