missing translation for 'onlineSavingsMsg'
Learn More

Draeger X-am™ 5100 Single-gas Detector

The Draeger X-am™ 5100 portable single-gas detector, customised to reactive gases ensures you use the safest methods possible for handling HF, HCl, H2O2 or hydrazine.

Brand:  Draeger 3361009

Product Code. 15880115

  • 2245.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • For personal air monitoring
  • Detection of HF, HCI, H2O2 or hydrazine
  • Ergonomic shape
  • Features lightweight smartphone form for greatest level of comfort
  • Two-button control panel and simple menu navigation for intuitive use
  • Continuous measurement - gives visual (360°), acoustic an vibration warnings
  • Direct gas entry ensures the sensors react quickly
  • Precise measurement data avoiding gas adsorption in the device housing
  • Robust design with integrated rubber boot and shock-resistant sensors offer extra safety in case of impact or vibrations
  • Resistant to electromagnetic irradiation
  • Can be combined with the X-zone 5500 for flexible area monitoring
Specifications

Specifications

Visual(360°), Audible (multi-tone >90 dB (at 30 cm; 1 ft.), Vibration)
Data logger can be read out via infrared interface; >1,000 h (at a recording interval of 1 value / minute)
For Manufacturing Petrochemical Products, Aseptic Packing, or Handling of Rocket Fuel; for Handling HF, HCl, H2O2 or Hydrazine
700 to 1,300 Mbar
Ambient -20 to +50 deg;C
CE marked, Electromagnetic compatibility - Directive 2014/30/EU; Explosion protection - Directive 2014/34/EU
48 x 130 x 61 mm
10 ro 90% r.h.
IP54
220 to 250 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.