Learn More
ABEBA™ X-Light Black Safety Trainers S3 ESD
Black X-light safety trainer with silver ion lining to reduce odor, replaceable active comfort insole, steel toe cap, textile antipuncture midsole, PU slip resistant outsole
Brand:  ABEBA™ 7131874/42
99.60 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 17171585
  • 126.00 € / 1 pair
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Lightweight, comfortable safety trainer
  • Comply to EN ISO 20345: 2012, S3 SRC
  • ESD dissipative according EN 61340 class climatic 1
  • Slip resistant Mono PU outsole SRC
  • Steel toe cap (200 J)
  • Textile antipuncture midsole
  • Black smooth leather
  • Silver ions lining
  • Replaceable active comfort insole
Specifications
Specifications
Black
42
Comply to EN 20345: 2012 S3 SRC, ESD according EN 61340
Low-cut
S3 SRC ESD
Leather
Steel
Unisex
Water Resistant