Learn More
ABEBA™ X-Light White Occupational Clogs OB
ABEBA™ X-Light White Occupational Clogs OB with silver ions lining to reduce odor, a replaceable Active Comfort insole, and a fixed heel strap.
Brand:  ABEBA™ 7131130/48
 View more versions of this product
Product Code. 16308391
  • 97.70 € / 1 pair
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Lightweight, comfort, hygiene
  • Comply to EN ISO 20347 : 2012, OB SRC
  • ESD dissipative according to EN 61340 Climate class 1
  • Slip resistant Mono PU outsole SRC
  • Smooth leather
  • Silver ions lining
  • Fixed heel strap
  • Replaceable Active Comfort insole
  • Sanitized treatment
Specifications
Specifications
CE EN ISO 20347:2012 OB, FO, SRC, ESD
Breathable lining with Silver ions
48
Polyurethane
1 Pair
Water
White
Leather
Slip and abrasion resistant, shock absorbing, ESD dissipative
Clogs
CE EN ISO 20347:2012, DIN EN 61340