Learn More
Draeger Safety X-Plore™ 4740 TPE Body
Draeger Safety X-Plore™ 4740 TPE Body is a half mask that is robust but also offers excellent comfort and a tight fit to the face.
Brand: Draeger Safety R55876
This item is not returnable.
View return policy
Description
- Designed to protect yourself from dangerous gases, vapors, and particles
- Exhalation valve is positioned so moisture escapes the mask and a dry atmosphere is maintained
- Hard mask body is made from durable PP plastic
- Remove your mask without having to take off your safety hat using the drop-down strap system
Specifications
CE certified (EN 140), AS/NZS | |
Medium/Large |