Learn More
Draeger Safety™ X-Plore 6300 Full Face Mask
The X-Plore 6300 Full Face Mask is for the price conscious user who demands quality and wearing comfort. The double layered face seal for triple sealing action ensures a comfortable and tight fit. The visor is made of highly scratch resistant plexiglass (PMMA). Without neck strap.
Brand:  Draeger Safety™ R55800
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10679274
  • 154.00 € / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Secure and comfortable fit
  • Mask body made from durable and skin-friendly EPDM
  • Double-layer face seal and triple sealing edges for a secure and comfortable seal for every face type
  • 180° panoramic view
  • Scratch-proof and distortion-free PMMA plexiglas visor
  • Efficient ventilation system - separate inner mask prevents the lens from fogging
  • 5-point harness - donned easily and quickly
  • Easy maintenance and logistics
  • Universal fit
  • Barcode on the inside simplifies recording and maintaining of the mask inventory
  • Versatile use
  • Equipped with a Rd40 connection in accordance with EN 148-1 and can be combined with Rd40 filter range
Specifications
Specifications
CE, EN 136
Standard thread
Full Face Mask
Personal
Anti-fog, Anti-scratch
X-plore™ 6300
1 Ea.
EN 136 Class 2 (CE mark), NIOSH, AS/NZS
Black
Yes
Adjustable 5-point
Polycarbonate
No