Learn More
uvex X-Tended Support BOA™ Black/Yellow 65682 Low Shoe - Width 11
Uvex 1 safety shoes protect and support the natural movement of the human body by reducing pressure and strain; protection classes S1 P and S3
155.00 €
Specifications
Color Black, Yellow
Material Microvelour
Sole Material Dual-density polyurethane (PU/PU)
Toe Material Composite
Disposable Reusable
View More Specs
Products 18
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17262291
View Documents
uvex
6568235
35
155.00 €
1 pair
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17272291
View Documents
uvex
6568236
36
155.00 €
1 pair
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17282291
View Documents
uvex
6568237
37
155.00 €
1 pair
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17292291
View Documents
uvex
6568238
38
155.00 €
1 pair
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17202301
View Documents
uvex
6568239
39
155.00 €
1 pair
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17212301
View Documents
uvex
6568240
40
155.00 €
1 pair
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17222301
View Documents
uvex
6568241
41
155.00 €
1 pair
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17232301
View Documents
uvex
6568242
42
155.00 €
1 pair
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17242301
View Documents
uvex
6568243
43
155.00 €
1 pair
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17252301
View Documents
uvex
6568244
44
155.00 €
1 pair
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17262301
View Documents
uvex
6568245
45
155.00 €
1 pair
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17272301
View Documents
uvex
6568246
46
155.00 €
1 pair
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17282301
View Documents
uvex
6568247
47
155.00 €
1 pair
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17292301
View Documents
uvex
6568248
48
155.00 €
1 pair
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17202311
View Documents
uvex
6568249
49
155.00 €
1 pair
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17212311
View Documents
uvex
6568250
50
155.00 €
1 pair
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17222311
View Documents
uvex
6568251
51
155.00 €
1 pair
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17232311
View Documents
uvex
6568252
52
155.00 €
1 pair
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
Description
Description
Uvex 1 X-Tended Support Shoe 6568.2 with BOA™ Fit System S1 P SRC
  • Sporty, multifunctional safety shoe
  • Lightweight and flexible
  • Latest generation of 100% metal-free Uvex Xenova™ toe cap
  • Compact design, anatomical shape, good side stability, no thermal conductivity and anti-magnetic properties
  • Penetration-resistant, non-metallic midsole in accordance with the latest safety standards, does not restrict flexibility
  • Made with new developed technology for extended wearing comfort
  • Uvex Climazone technology for optimum wearer comfort
  • Breathable, perforated, high-tech microvelour upper material
  • Distance mesh lining provides optimum breathability
  • Made from synthetic materials, suitable for people allergic to chrome
  • BOA™ Fit System Dial for custom comfort that's consistent all day
  • Virtually seamless design to eliminate pressure points
  • Removable comfort insole, antistatic, with moisture control system and additional shock absorption in heel and at front of foot
  • Soft, padded collar and tongue
  • Medial and lateral heel support for foot through especially shaped side frame to protect against ankle injuries, for guidance and impact protection
  • Outstanding dual density polyurethane sole unit with best shock absorption characteristics and superb slip resistance
    • Free of lacquer wetting inhibitors such as silicones, phthalates, etc.
  • Fulfills ESD requirements: resistance to ground Kleiner 35 megohms
  • Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
  • Article No. 6568.2
  • Standard EN ISO 20345:2011 S1 P SRC
  • Sizes: 35 to 52
Specifications
Specifications
Black, Yellow
Dual-density polyurethane (PU/PU)
Reusable
Unisex
Uvex 1
No
SRC
Microvelour
Composite
EN ISO 20345:2011
Standard Width
Yes
S1
ESD Safety Shoe
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
uvex X-Tended Support BOA™ Black/Yellow 65682 Low Shoe - Width 11