missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ X1 Side-Mount Oval Polypropylene Cupsink

The X1 Side-Mount Oval Polypropylene Cupsink mounts on the side wall of Protector XL Floor-Mounted Hoods.

Brand:  Labconco™ 4852600

Product Code. 13433918

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Spare part
  • 1.5 in. NPSM Thread
  • Requires 1.875 in. dia. drill hole in liner
Specifications

Specifications

Polypropylene
Oval
15.2 cm
7.6 cm
1.8 kg
Cupsink
6 in.
3 in.
4 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Labconco™ X1 Side-Mount Oval Polypropylene Cupsink > Shape: Oval; Material: Polypropylene

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.