missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDPE Wide Neck 5L Bottle
Click to view available options
Capacity (Metric):
5 L
Description
- Supplied with tamper evident polypropylene cap
- Moulded in carrying/pouring handle
- Unapproved
- Color: Clear
Specifications
Specifications
Capacity (Metric) | 5 L |
Color | Clear |
Diameter (Metric) Outer Neck | 45 mm |
Material | HDPE |
Quantity | 1 |
Width (Metric) | 170mm |
Certifications/Compliance | Tested and certified to meet UN standards for the safe transportation of hazardous materials |
Height (Metric) | 330 mm |
Closure Material | Polypropylene |
Closure Type | Tamper-evident |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'